Order Entry
India
ContactUsLinkComponent
Designed for synchronous heating and stirring, Has consistent stirring on all positions, Homogeneous temperature distribution across , the heating plate, Silicone surface for anti-skid. Prominently used in chemical synthesis, pharmaceuticals, physica 1 * 1 items
Catalog # 442-1261
Supplier:  VWR Collection
Designed for synchronous heating and stirring, Has consistent stirring on all positions, Homogeneous temperature distribution across , the heating plate, Silicone surface for anti-skid. Prominently used in chemical synthesis, pharmaceuticals, physica 1 * 1 items
Catalog # 442-1261
Supplier:  VWR Collection
Frequently Bought Together