Order Entry
India
ContactUsLinkComponent
High tech motor causes low working noise, Produces low vibration with advanced shock absorbs, Compact design and can carry higher vo lume of 2 ml x 8 places, Easy replacement of rotor, Maximum relative centrifugal force upto 3260G, Fast Stopping in 1 1 * 1 items
Catalog # 521-2573
Supplier:  VWR Collection
High tech motor causes low working noise, Produces low vibration with advanced shock absorbs, Compact design and can carry higher vo lume of 2 ml x 8 places, Easy replacement of rotor, Maximum relative centrifugal force upto 3260G, Fast Stopping in 1 1 * 1 items
Catalog # 521-2573
Supplier:  VWR Collection
Frequently Bought Together