Order Entry
India
ContactUsLinkComponent
TransduceIT Reagent is an aqueous solution of hexadimethrine bromide, a cationic polymer, that is shown to greatly enhance retroviral transduction and transgene expression in both dividing and quiescent mammalian cells. Provided as 1mL vial. 1 * 1 mL
Catalog # MIRUMIR6620
Supplier:  MIRUS
TransduceIT Reagent is an aqueous solution of hexadimethrine bromide, a cationic polymer, that is shown to greatly enhance retroviral transduction and transgene expression in both dividing and quiescent mammalian cells. Provided as 1mL vial. 1 * 1 mL
Catalog # MIRUMIR6620
Supplier:  MIRUS
Frequently Bought Together