FEP BAG Outside Dimensions, cm 9.7 x 17.3 Inside Dimensions, cm 8.6 x 11.2 Inside Surface Area, cm² 193 Permeability O2 (gas) cc/day @ 37°C : 59 CO2 (gas) cc/day @ 37°C : 137N2 (gas) cc/day @ 37°C : 22H2O (liquid) cc/day @ 37°C : 0.01
Catalog # VERNLP-0100
Supplier: Saint Gobain Life Sciences