Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Transplant Medium (CMRL 1066) without phenol red 1 * 500 mL
Catalog # MDTC99-663-CV
Supplier:
Corning
Expand image
Transplant Medium (CMRL 1066) without phenol red 1 * 500 mL
Catalog # MDTC99-663-CV
Supplier:
Corning
Related Information
Documentation
Corning Cell culture media and reagents - Product selection guide
Documentation
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection