Order Entry
India
ContactUsLinkComponent
Chart Recorder, Inkless VWR Collection Chest Freezers CI, Field Installed Option. This uses a pen with no ink where the marking is created by pressure. 1 * 1 items
Catalog # 471-1149
Supplier:  VWR Collection
Chart Recorder, Inkless VWR Collection Chest Freezers CI, Field Installed Option. This uses a pen with no ink where the marking is created by pressure. 1 * 1 items
Catalog # 471-1149
Supplier:  VWR Collection
Frequently Bought Together