Buffers, pH reference standards, BDH®
Buffer, pH reference standard, 10,00, ±0,01, Colourless, 25 °C, Cubitainer®, 20 L
BDHB5082-20L
Buffers are available as colourless and coloured solutions.
: ISO 17025 and traceable to NIST.
- Pack type:Cubitainer®
- Pk:20 L
- Reference temperature:25 °C
- Colour:Colourless
- Description:Buffer, pH reference standard
- pH accuracy:±0,01
- pH value:10,00



