Order Entry
India
ContactUsLinkComponent
The CHOgro Expression System is an optimized platform for transient, high titer protein production in suspension CHO cells. This sys tem consists of CHOgro Expression Medium, L-Glutamine and Poloxamer 188 medium supplements, TransIT-PRO Transfection 1 * 1 KIT
  MIRUMIR6260
 :  MIRUS
The CHOgro Expression System is an optimized platform for transient, high titer protein production in suspension CHO cells. This sys tem consists of CHOgro Expression Medium, L-Glutamine and Poloxamer 188 medium supplements, TransIT-PRO Transfection 1 * 1 KIT
  MIRUMIR6260
 :  MIRUS
Frequently Bought Together