Order Entry
India
ContactUsLinkComponent
L-Glutamine is an essential amino acid added as a carbon and energy source to mammalian and insect cell culture media. Optimal cell performance usually requires 2-4 mM. L-Glutamine gradually degrades in stock solutions and liquid media and should be 1 * 100 mL
Catalog # MIRUMIR6240
Supplier:  MIRUS
L-Glutamine is an essential amino acid added as a carbon and energy source to mammalian and insect cell culture media. Optimal cell performance usually requires 2-4 mM. L-Glutamine gradually degrades in stock solutions and liquid media and should be 1 * 100 mL
Catalog # MIRUMIR6240
Supplier:  MIRUS
Frequently Bought Together