Order Entry
India
ContactUsLinkComponent
CHOgro Complex Formation Solution is a chemically defined and animal origin-free medium. It is for exclusive use with the TransIT-PR O Transfection Reagent and the CHOgro Expression System, a platform designed for high and reproducible protein yield 1 * 100 mL
Catalog # MIRUMIR6210
Supplier:  MIRUS
CHOgro Complex Formation Solution is a chemically defined and animal origin-free medium. It is for exclusive use with the TransIT-PR O Transfection Reagent and the CHOgro Expression System, a platform designed for high and reproducible protein yield 1 * 100 mL
Catalog # MIRUMIR6210
Supplier:  MIRUS
Frequently Bought Together