Order Entry
India
Orders LinkContactUsLinkComponent
Baculovirus expression system, flashBAC™
flashBAC™ ULTRA baculovirus expression system, 24 reactions, 1 KIT
Catalog # MIRUMIR6140
Supplier:   MIRUS
Baculovirus expression system, flashBAC™
Catalog # MIRUMIR6140
Supplier:  MIRUS
CAS Number:  
Baculovirus expression system, flashBAC™
flashBAC™ ULTRA baculovirus expression system, 24 reactions, 1 KIT
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
The flashBAC™ Baculovirus Expression System is designed for high yield protein production in insect and mammalian cells.
  • Single-step baculovirus generation in insect cells, no plaque purification required
  • Ready to use with most transfer vectors
  • High recombinant protein yields
This system is a streamlined platform for the production of recombinant baculoviruses. Genetic optimisation of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    flashBAC™ ULTRA baculovirus expression system, 24 reactions
Specifications