Baculovirus expression system, flashBAC™
flashBAC™ Baculovirus expression system, 5 reactions, 1 KIT
MIRUMIR6115
: MIRUS
The flashBAC™ Baculovirus Expression System is designed for high yield protein production in insect and mammalian cells.
- Single-step baculovirus generation in insect cells, no plaque purification required
- Ready to use with most transfer vectors
- High recombinant protein yields
This system is a streamlined platform for the production of recombinant baculoviruses. Genetic optimisation of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification.
- Pk:1 KIT
- Description:flashBAC™ Baculovirus expression system, 5 reactions