Order Entry
India
Orders LinkContactUsLinkComponent
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol red, Powder, 10 L
Catalog # 90-050-PB
Supplier:   Corning
Medium 199 (Mod.), Corning®
Catalog # 90-050-PB
Supplier:  Corning
CAS Number:  
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), Corning®
Medium 199 (Mod.), powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol red, Powder, 10 L
CAS Number:  
Price
Rs.18,885.04
Log in for availability
1 * 10 L
Product Details & Documents
Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.
Specifications
Specifications
  • Media Format:
    Powder
  • Pk:
    10 L
  • Description:
    Medium 199 (Mod.), powder [+] Earle's salts [-] sodium bicarbonate, L-glutamine, phenol red
Specifications