Order Entry
India
Orders LinkContactUsLinkComponent
VWR® VistaVision, Microscope slides, colour-coded

VWR® VistaVision, Microscope slides, colour-coded

Microscopy slides, premium, square corners, White, 1,0 mm, 75×25 mm
Catalog # VWRU16004-382
Supplier:   VWR Collection
VWR® VistaVision, Microscope slides, colour-coded
Catalog # VWRU16004-382
Supplier:  VWR Collection
VWR® VistaVision, Microscope slides, colour-coded

Microscopy slides, premium, square corners, White, 1,0 mm, 75×25 mm

Price
Inquire For Price
Log in for availability
1 * 72 items
Product Details & Documents

Bright printed colours of these microscope slides provide an ideal marking area and enable easy identification. The ultra-thin colour frosted end facilitates processing on automated systems.

  • Manufactured using high quality, white soda lime glass
  • Pre-cleaned and ready for use
  • Autoclavable
  • Colour coded ends facilitate sorting and identification
  • Uniform size and thickness

Certifications: ISO 8037/1

Specifications
  • Colour:
    White
  • Description:
    Microscopy slides, premium, square corners
  • L×W:
    75×25 mm
  • Seq:
    02
  • Thickness:
    1,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.