Buffers, pH reference standards, BDH®
Buffer, pH reference standard, 4,00, ±0,01, Colourless, 25 °C, Poly bottle, 4 L
BDHB5026-4L
Buffers are available as colourless and coloured solutions.
: ISO 17025 and traceable to NIST.
- Pack type:Poly bottle
- Pk:4 L
- Reference temperature:25 °C
- Colour:Colourless
- Description:Buffer, pH reference standard
- pH accuracy:±0,01
- pH value:4,00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



