Buffers, pH reference standards, BDH®
Buffer, pH reference standard, 4,00, ±0,01, Red, 25 °C, Cubitainer®, 20 L
Catalog # BDHB0198-20L
Supplier: BDH US PRODUCED
Jump to:
Product Details & Documents
Buffers are available as colourless and coloured solutions.
Certifications: ISO 17025 and traceable to NIST.
Specifications
Specifications
- Pack type:Cubitainer®
- Pk:20 L
- Reference temperature:25 °C
- Colour:Red
- Description:Buffer, pH reference standard
- pH accuracy:±0,01
- pH value:4,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



