Buffers, pH reference standards, BDH®
Buffer, pH reference standard, 4,00, ±0,01, Red, 25 °C, Poly drum, 208 L
BDHB0196-55GL
Buffers are available as colourless and coloured solutions.
: ISO 17025 and traceable to NIST.
- Pack type:Poly drum
- Pk:208 L
- Reference temperature:25 °C
- Colour:Red
- Description:Buffer, pH reference standard
- pH accuracy:±0,01
- pH value:4,00
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



