Order Entry
India
Orders LinkContactUsLinkComponent
Buffers, pH reference standards, BDH®

Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 7,00, ±0,01, Yellow, 25 °C, Cubitainer®, 20 L
Catalog # BDHB0194-20L
Supplier:   BDH US PRODUCED
Buffers, pH reference standards, BDH®
Catalog # BDHB0194-20L
Supplier:  BDH US PRODUCED
CAS Number:  
Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 7,00, ±0,01, Yellow, 25 °C, Cubitainer®, 20 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents

Buffers are available as colourless and coloured solutions.

Certifications: ISO 17025 and traceable to NIST.

Documents

Specifications

Specifications

  • Pack type:
    Cubitainer®
  • Pk:
    20 L
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • Description:
    Buffer, pH reference standard
  • pH accuracy:
    ±0,01
  • pH value:
    7,00

Specifications