Order Entry
India
ContactUsLinkComponent
Conical Bottom Centrifuge Tube 15ml - Non Sterile. Certified Dnase, Rnase and Pyrogen Free. Resin Confirming to US FDA and USP Class VI Requiremets. Leak proof tubes, have printed vloume graduation and writting area along the length of the tube. Perf 1 * 500 items
Catalog # 734-2669
Supplier:  VWR Collection
Conical Bottom Centrifuge Tube 15ml - Non Sterile. Certified Dnase, Rnase and Pyrogen Free. Resin Confirming to US FDA and USP Class VI Requiremets. Leak proof tubes, have printed vloume graduation and writting area along the length of the tube. Perf 1 * 500 items
Catalog # 734-2669
Supplier:  VWR Collection
Frequently Bought Together