Order Entry
India
Orders LinkContactUsLinkComponent
 

Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 12,00, ±0,02, Colourless, 25 °C, Poly bottle, 500 mL
Catalog # BDHB5086-500ML
Supplier:   BDH US PRODUCED
Buffers, pH reference standards, BDH®
Catalog # BDHB5086-500ML
Supplier:  BDH US PRODUCED
CAS Number:  
Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 12,00, ±0,02, Colourless, 25 °C, Poly bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

Buffers are available as colourless and coloured solutions.

 : ISO 17025 and traceable to NIST.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • Description:
    Buffer, pH reference standard
  • pH accuracy:
    ±0,02
  • pH value:
    12,00

Specifications