Order Entry
India
Orders LinkContactUsLinkComponent
VWR®, Sample library tubes

VWR®, Sample library tubes

8-well tube strips, Clear
Catalog # VWRU82006-700
Supplier:   VWR Collection
VWR®, Sample library tubes
Catalog # VWRU82006-700
Supplier:  VWR Collection
VWR®, Sample library tubes
VWR®, Sample library tubes

8-well tube strips, Clear

Price
Inquire For Price
Log in for availability
1 * 960 items
Product Details & Documents

PP, clear.

  • Autoclavable and freezable (–80 °C)
  • Printed with an alpha numeric grid that helps to find a sample easily

These 1,2 ml sample library tubes are supplied in a 96 tube format for storage of specimens. The rack is designed to meet SBS standards and can be used in robotic applications.

Delivery information: Supplied in 8×12 racks with clear, snug fit lid.

Specifications
  • Colour:
    Clear
  • Description:
    8-well tube strips
  • Seq:
    05
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • 12-well closure strips, sterile
    12-well closure strips, sterile
    VWRU93000-048
    Log in for availability
    1 * 960 items