Rs.0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
India
ContactUsLinkComponent
Mineral Stabilizer Solution, 500 ml, used in Ammonia (8038) and TKN (8075) Method for soils, sludges and wastewater samples 1 * 500 mL
HACH2376649
:
HACH
Mineral Stabilizer Solution, 500 ml, used in Ammonia (8038) and TKN (8075) Method for soils, sludges and wastewater samples 1 * 500 mL
HACH2376649
:
HACH
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Powder pillows, aluminium
: HACH
VWR® Facial Tissues
: VWR Collection
Test strips, nitrate/nitrite, Dosatest
: VWR Chemicals
Powder pillows, silica
: HACH
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals