Order Entry
India
Orders LinkContactUsLinkComponent
Buffers, pH reference standards, BDH®

Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 9,18, ±0,01, Colourless, 25 °C, Poly bottle, 500 mL
Catalog # BDHB5066-500ML
Supplier:   BDH US PRODUCED
Buffers, pH reference standards, BDH®
Catalog # BDHB5066-500ML
Supplier:  BDH US PRODUCED
CAS Number:  
Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 9,18, ±0,01, Colourless, 25 °C, Poly bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

Buffers are available as colourless and coloured solutions.

Certifications: ISO 17025 and traceable to NIST.

Documents

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • Description:
    Buffer, pH reference standard
  • pH accuracy:
    ±0,01
  • pH value:
    9,18

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.