Order Entry
India
ContactUsLinkComponent
Chloride Reagent Set, Low Range, DT, 100 tests, MR 10 - 100 mg/L as Cl-, HACH Digital Titrator Method 8207, Chloride - Silver Nitr ate Method 1 * 1 items
Catalog # HACH2444100
Supplier:  HACH
undefined
Chloride Reagent Set, Low Range, DT, 100 tests, MR 10 - 100 mg/L as Cl-, HACH Digital Titrator Method 8207, Chloride - Silver Nitr ate Method 1 * 1 items
Catalog # HACH2444100
Supplier:  HACH
Frequently Bought Together