Order Entry
India
ContactUsLinkComponent
Vaccine & Icepack Freezer, Voltage: 220/50 input power (W): 134, Temp: -15-25 , capacity: 116L, Weight: 58 kg, Basket: 2 Ext Dim (WxDxH)mm: 669*628*818 Int Dim (WxDxH)mm: 497*457*650 1 * 1 items
Catalog # HAIEHBD-116
Vaccine & Icepack Freezer, Voltage: 220/50 input power (W): 134, Temp: -15-25 , capacity: 116L, Weight: 58 kg, Basket: 2 Ext Dim (WxDxH)mm: 669*628*818 Int Dim (WxDxH)mm: 497*457*650 1 * 1 items
Catalog # HAIEHBD-116
Frequently Bought Together