Order Entry
India
ContactUsLinkComponent
RapidVap N2 48-PLACE BLOCK for 20 mm TUBES 20 x 150 mm OD Sample Tube PTFE-Coated Aluminium Block For use with RapidVap N2/48 Evaporation System. Block tube capacity: 48. Sample tube size: 20 x 150 mm OD tubes, up to 35 ml. Sample volume: 26 ml. Actu 1 * 1 items
Catalog # LBCP7482400
Supplier:  LABCONCO
RapidVap N2 48-PLACE BLOCK for 20 mm TUBES 20 x 150 mm OD Sample Tube PTFE-Coated Aluminium Block For use with RapidVap N2/48 Evaporation System. Block tube capacity: 48. Sample tube size: 20 x 150 mm OD tubes, up to 35 ml. Sample volume: 26 ml. Actu 1 * 1 items
Catalog # LBCP7482400
Supplier:  LABCONCO
Frequently Bought Together