Transfection kit and reagent, TransIT-PRO®
TransIT-PRO® transfection kit, 1,0 ml, with 1,5 ml PRO boost reagent, 1 mL
MIRUMIR5700
: MIRUS
TransIT-PRO® transfection kit and reagents are ideal for biotherapeutic protein production in suspension CHO and 293 cells.
- High protein yield in suspension CHO and 293 cells, including CHO-S and Expi293F™ cells
- Compatible with multiple CHO media formulations; the same media to be used for both transient and stable expression
- Decreases time to produce usable protein by maximising target protein yields through transient transfection
- Simple workflow enables reproducible protein expression with minimal optimisation
: Kit contains TransIT-PRO® transfection reagent and PRO boost reagent. TransIT-PRO® transfection reagent also available separately.
- Pk:1 mL
- Description:TransIT-PRO® transfection kit, 1,0 ml, with 1,5 ml PRO boost reagent
- Cat. No.:MIRUMIR5700
- Cell specific:293, CHO
- DNA:Yes
- Virus production:Yes
- Bioproduction:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



