Order Entry
India
Orders LinkContactUsLinkComponent
VWR®, Microscope slide boxes
VWR®, Microscope slide boxes
Blue, Plastic, 25, 141×88×35 mm
Catalog # VWRU82024-614
Supplier:   VWR Collection
VWR®, Microscope slide boxes
Catalog # VWRU82024-614
Supplier:  VWR Collection
VWR®, Microscope slide boxes
VWR®, Microscope slide boxes
Blue, Plastic, 25, 141×88×35 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
These boxes made of PP are designed for the storage or transport of either 25 or 100 microscopy slides.
  • Boxes are unlined
  • Hinged lids are held securely closed by latch locks
  • Colour-coded for ease in sorting and selection
Convenient, space-saving design with stacking feet maximises stability when boxes are stored atop one another.
Specifications
  • No. of slides:
    25
  • Colour:
    Blue
  • Material:
    Plastic
  • Seq:
    02
  • W×D×H:
    141×88×35 mm