VWR®, Microscope slide boxes
White, Plastic, 25, 141×88×35 mm
Catalog # VWRU82024-612
Supplier: VWR Collection
Jump to:
Product Details & Documents
These boxes made of PP are designed for the storage or transport of either 25 or 100 microscopy slides.
- Boxes are unlined
- Hinged lids are held securely closed by latch locks
- Colour-coded for ease in sorting and selection
Convenient, space-saving design with stacking feet maximises stability when boxes are stored atop one another.
Specifications
- No. of slides:25
- Colour:White
- Material:Plastic
- Seq:01
- W×D×H:141×88×35 mm