Order Entry
India
ContactUsLinkComponent
S-70 controller For BSS-3000, for liquid phase, supplied with connection tools to tank, Electronic control module, for monitoring temperature and level and automatic filling of the vessel when connected to LN2-supply. Supplied with 3 different power 1 * 1 items
Catalog # 478-0330
Supplier:  VWR Collection
S-70 controller For BSS-3000, for liquid phase, supplied with connection tools to tank, Electronic control module, for monitoring temperature and level and automatic filling of the vessel when connected to LN2-supply. Supplied with 3 different power 1 * 1 items
Catalog # 478-0330
Supplier:  VWR Collection
Frequently Bought Together