Accessories for heating circulators
Stainless steel/PTFE coated PT100 sensor with 3 m cable, 300×6 mm, CF31, CF41, F25-HE, F25-HL, F25-ME
Catalog # JULA8981015
Supplier: JULABO GMBH
Jump to:
Product Details & Documents
Specifications
- Description:Stainless steel/PTFE coated PT100 sensor with 3 m cable, 300×6 mm
- Seq:20
- For:CF31, CF41, F25-HE, F25-HL, F25-ME



