VWR®, Microscope slide boxes
Red, Plastic, 100, 208×175×34 mm
VWRU82024-610
These boxes made of PP are designed for the storage or transport of either 25 or 100 microscopy slides.
- Boxes are unlined
- Hinged lids are held securely closed by latch locks
- Colour-coded for ease in sorting and selection
Convenient, space-saving design with stacking feet maximises stability when boxes are stored atop one another.
- No. of slides:100
- Colour:Red
- Material:Plastic
- Seq:06
- W×D×H:208×175×34 mm