Order Entry
India
Orders LinkContactUsLinkComponent
Dnase
Dnase
200 Tests
Catalog # E1091-02
Supplier:   OMEGA BIOTEK
Dnase
Catalog # E1091-02
Supplier:  OMEGA BIOTEK
CAS Number:  
Dnase
200 Tests
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 200 Tests
Product Details & Documents
For DNase digestion during RNA purification. The RNase-free DNase set provides a rapid and efficient on-column digestion of DNA during RNA purification from a wide variety of sample sources using E.Z.N.A.® RNA kits. The DNases can be efficiently removed in subsequent steps. Generally, DNase digestion is not required for RNA purified with E.Z.N.A.® RNA kits since the silica-membrance, spin-column technology efficiently removes the majority of the DNA without DNase treatment. However, complete DNA removal may be necessary for certain RNA applications that are sensitive to very small amounts of DNA.
  • RNase-free
Specifications
Specifications
  • Pk:
    200 Tests
  • Cat. No.:
    E1091-02
  • Enzyme name:
    Dnase