Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LID FOLDING FOR NLK APPARATUS 1 * 1 items
Catalog # KGWA11217
Supplier:
KGW
Expand image
LID FOLDING FOR NLK APPARATUS 1 * 1 items
Catalog # KGWA11217
Supplier:
KGW
Frequently Bought Together
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: METTLER TOLEDO
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
VWR®, Hand Tally Counter
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK