Order Entry
India
Orders LinkContactUsLinkComponent
VWR®, Zero Air Generators

VWR®, Zero Air Generators

18 l/min, 280×330×304 mm
Catalog # VWRU26000-026
Supplier:   VWR Collection
VWR®, Zero Air Generators
Catalog # VWRU26000-026
Supplier:  VWR Collection
VWR®, Zero Air Generators
VWR®, Zero Air Generators

18 l/min, 280×330×304 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Designed as a compact, hazard free, lower cost alternative to the user of expensive high pressure gas cylinders in the laboratory. Replacing cylinders with a gas generator brings advantages : increased safety, greater convenience, reduced cost and higher purity gas for more accurate analysis.

  • Produces zero air from compressed air
  • Ensures a flat and consistent base line
  • Short payback period

Purity: <0,05 ppm THC.

Inlet pressure 2,8 to 8,6 bar.

Inlet/outlet port size: ¹/₄" NPT female.

Specifications
  • Flow rate:
    18 l/min
  • Seq:
    02
  • W×D×H:
    280×330×304 mm