Order Entry
India
Orders LinkContactUsLinkComponent
E.Z.N.A.® Soil DNA kit

E.Z.N.A.® Soil DNA kit

E.Z.N.A.® Soil DNA Mini Kit, 200 Tests
Catalog # D5625-02
Supplier:   OMEGA BIOTEK
E.Z.N.A.® Soil DNA kit
Catalog # D5625-02
Supplier:  OMEGA BIOTEK
CAS Number:  
E.Z.N.A.® Soil DNA kit
E.Z.N.A.® Soil DNA kit

E.Z.N.A.® Soil DNA Mini Kit, 200 Tests

CAS Number:  
Price
Rs.198,956.36
Log in for availability
1 * 200 Tests
Product Details & Documents

The E.Z.N.A.® soil DNA kit is formulated to isolate high purity cellular DNA from soil samples typically containing humic acid and inhibitors of PCR. This kit has been successfully used to isolate DNA from tough-to-lyse bacteria, fungi, and algae that inhabit a range of samples including clay, sandy, peaty, chalky, and loamy soil samples. Isolated DNA can be used for most downstream applications including PCR, southern blotting, and SNP analysis.

  • DNA isolation in 1 hour
  • Optimised buffers guarantee pure DNA every time
  • Purified DNA suitable for most downstream applications
Specifications

Specifications

  • Pk:
    200 Tests
  • Description:
    E.Z.N.A.® Soil DNA Mini Kit
  • Cat. No.:
    D5625-02
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)
  • Description:
    E.Z.N.A.® Soil DNA Mini Kit
  • Sample type:
    Soil
  • Sample size:
    1 g

Specifications

Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • cHTR reagent, 50 ml
    cHTR reagent, 50 ml
    CHTR-50
    50 mL
    Log in for availability
    1 * 50 mL - Rs.55,683.86

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • P2 buffer
    P2 buffer
    PD076
    60 mL
    Log in for availability
    1 * 60 mL - Rs.19,452.42

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Elution buffer
    Elution buffer
    PDR048
    100 mL
    Log in for availability
    1 * 100 mL

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.