Order Entry
India
Orders LinkContactUsLinkComponent
Plasmid delivery controls, Label IT®
Plasmid delivery controls, Label IT®
Label IT® plasmid delivery control, fluorescein, 10 µG
Catalog # MIRUMIR7906
Supplier:   MIRUS
Plasmid delivery controls, Label IT®
Catalog # MIRUMIR7906
Supplier:  MIRUS
CAS Number:  
Plasmid delivery controls, Label IT®
Label IT® plasmid delivery control, fluorescein, 10 µG
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 µG
Product Details & Documents
Fluorescently labelled plasmids ideal for in vitro and in vivo tracking.
  • For in vivo delivery
  • Easily visualise transfected cells and assess delivery efficiency using fluorescent microscopy
  • Suitable for co-delivery experiments with functional plasmids
Delivery information: Supplied as a ready to use 0,5 mg/ml concentrate.
Specifications
Specifications
  • Pk:
    10 µG
  • Description:
    Label IT® plasmid delivery control, fluorescein