Surface active cleaning agent, decon® 90
Decon 90®, Plastic bottle
Supplier: DECON LAB
Jump to:
Product Details & Documents
A surface active cleaning agent and/or radioactive decontaminant, for laboratory, medical and specialised industrial applications. A concentrate that is suitable for ‘manual’ cleaning, or for use in an ultrasonic cleaning bath.
- Biodegradable, phosphate-free and enzyme-free
- Bactericidal
- Highly alkaline >pH 13
Certifications: decon® 90 satisfies the criteria in BS EN 1040 1997 for bacterial activity.
Caution: Not suitable for domestic use nor suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate.
Specifications
- Description:Decon 90®
- Pack type:Plastic bottle
- Cat. No.:148-0325