Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Coenzyme A trilithium salt dihydrate BioChemica 1 * 100 mg
Catalog # A0813.0100
Supplier:
PanReac AppliChem
Expand image
Coenzyme A trilithium salt dihydrate BioChemica 1 * 100 mg
Catalog # A0813.0100
Supplier:
PanReac AppliChem
Frequently Bought Together
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals