Accessories for heating circulators
Stainless steel/PTFE coated PT100 sensor with 3 m cable, 600×6 mm, CF31, CF41, F25-HE, F25-HL, F25-ME
Catalog # JULA8981013
Supplier: JULABO GMBH
Jump to:
Product Details & Documents
Specifications
- Description:Stainless steel/PTFE coated PT100 sensor with 3 m cable, 600×6 mm
- Seq:21
- For:CF31, CF41, F25-HE, F25-HL, F25-ME
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



