Rs.0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
India
ContactUsLinkComponent
PHOSPHATE STANDARD SOLUTION 30MG/L 1 * 2,9 L
HACH1436703
:
HACH
PHOSPHATE STANDARD SOLUTION 30MG/L 1 * 2,9 L
HACH1436703
:
HACH
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals