Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Silica ULR Ampuled Reagent Set, 40 tests, MR 3 - 1000 ?g/L as SiO2, HACH Method 8282, Heteropoly Blue Method 1 * 40 Tests
Catalog # HACH2581400
Supplier:
HACH
Expand image
Silica ULR Ampuled Reagent Set, 40 tests, MR 3 - 1000 ?g/L as SiO2, HACH Method 8282, Heteropoly Blue Method 1 * 40 Tests
Catalog # HACH2581400
Supplier:
HACH
Related Information
Documentation
Hach Chemistries, Reagents and Standards
Documentation
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals