Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SPADNS Fluoride Reagent Solution, 100 ml, 25 tests, MR 0.02 - 2.00 mg/L F, HACH Method 8029, SPADNS Reagent 1 * 100 mL
Catalog # HACH44442
Supplier:
HACH
Expand image
SPADNS Fluoride Reagent Solution, 100 ml, 25 tests, MR 0.02 - 2.00 mg/L F, HACH Method 8029, SPADNS Reagent 1 * 100 mL
Catalog # HACH44442
Supplier:
HACH
Frequently Bought Together
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals