Order Entry
India
Orders LinkContactUsLinkComponent
 

Samplers, Liquid-Sampler

Liquid-Sampler, open with thumb, 32×460 mm, 50 ml
  331-0032
 :   BURKLE
Samplers, Liquid-Sampler
  331-0032
 :  BURKLE
Samplers, Liquid-Sampler
Samplers, Liquid-Sampler

Liquid-Sampler, open with thumb, 32×460 mm, 50 ml

 
 
   
1 * 1 items
 

Material: V4A stainless steel (1.4404). Sampler for thin to viscous liquids.

  • Easy one handed operation
  • Discharge funnel for safe filling of sample bottles
 
  • Capacity:
    50 ml
  • Description:
    Liquid-Sampler, open with thumb, 32×460 mm