0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FILTER DISC CELLULOSE PAPER MODERATELY FAST DIAMETER-185MM 1 * 100 items
Catalog # 515-1228
Supplier:
Sartorius
Expand image
FILTER DISC CELLULOSE PAPER MODERATELY FAST DIAMETER-185MM 1 * 100 items
Catalog # 515-1228
Supplier:
Sartorius
Frequently Bought Together
VWR®, Syringe Filters
Supplier: VWR Collection
VWR® Weighing Paper, Grade 531
Supplier: VWR Collection
VWR®, Glass Fibre Filters
Supplier: VWR Collection
VWR®, Bottle-Top Vacuum Filtration Systems, PES
Supplier: VWR Collection
VWR®, Qualitative Filter Papers
Supplier: VWR Collection