Order Entry
India
Orders LinkContactUsLinkComponent
 

VWR®, Clamps

8 mm rod Ø, 8 - 30 mm, 125
  241-0013
VWR®, Clamps
  241-0013
VWR®, Clamps
VWR®, Clamps

8 mm rod Ø, 8 - 30 mm, 125

 
 
   
1 * 5 items
 

Laboral aluminium alloy.

  • Round jaws covered in vinyl to protect clamped items
  • One easy action knurled screw for securing
 
  • Spanning width:
    8 - 30 mm
  • Description:
    8 mm rod Ø
  • Length:
    125
  • Seq:
    01
Accessories

 

  •  
    Description
    For
     
     
  • 241-0150
    Protecting clamp for clamp jaws
    Clamps 241-0013
       
    1 * 10 items

 

  •  
    Description
    For
     
     
  • 241-0151
    Protecting clamp for clamp jaws
    Clamps 241-0020/241-0023
       
    1 * 10 items