Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Catalog # HAMI238217
Supplier:
HAMILTON BONADUZ
Certificates
Expand image
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Catalog # HAMI238217
Supplier:
HAMILTON BONADUZ
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
VWR®, Twist-Seal Bags
Supplier: VWR Collection
Bottles, narrow neck, square, PET, with tamper-proof closure
Supplier: BURKLE
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ