Order Entry
India
ContactUsLinkComponent
VWR® Microplate Washer for 96 Well Plates
VWR® Microplate Washer for 96 Well Plates
Supplier:  VWR Collection
New Product
Certificates
VWR® Microplate Washer for 96 Well Plates
Supplier:  VWR Collection
New Product
Specifications
  • Catalog No:
  • 000000000008120391
  • 000000000008123732
  • 000000000008120390
  • Wash heads:
  • 8 channel or 12 channel
  • Washing time:
  • 00:00 ~ 03:59 (mm:ss), rinse function time range; 00:00 ~ 99:00 (mm:ss), soak and shaking time range
  • Dispensing volume range:
  • 50 - 2000 µl
  • Display:
  • 4,3" LCD display
  • Program memory:
  • 100 programs
  • W×D×H:
  • 342,9×457,2×215,9 mm
  • Weight:
  • 11,34 kg