


Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
: Avantor
:7761-88-8
:
:AgNO₃
:MFCD00003414
:169.87 g/mol
:Ambient
:231-853-9
:3082
:9,III
:GHS06
:GHS08
:GHS09
- For Laboratory,Research,or Manufacturing Use:0.095 - 0.105
- Normality:0.095 - 0.105
- pH at 25°C:3.0 - 6.0
- Chloride (Cl):≤1 ppm
- Sulfate (SO₄):≤1 ppm
- Appearance:Passes Test
Frequently Bought Together