D-(-)-Sorbitol ≥91.0%, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
V045-09
: Avantor
:
- Pack type:PE pail, screw top
- Pk:3 kg
- Seq:4090
- For Laboratory,Research,or Manufacturing Use:= 91.0 %
- Assay (C₆H₁₄O₆):≥91.0 %
- Ash (sulfated):≤0.1 %
- Loss on Drying at 80°C (in vacuo):≤1.0 %
- Reducing Sugars:≤0.3 %
- Total Sugars:≤1.0 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO₄):≤0.01 %
- Arsenic (As):≤0.0003 %
- Heavy Metals (as Pb):≤0.001 %
:
:13,08797
:295 °C (1013 hPa)
:94…98 °C
:1,524 g/cm³ (20 °C)
:>100 °C
:182.17 g/mol
:Ambient
:200-061-5
:50-70-4
:98201-93-5
:HOH₂C(CH(OH))₄CH₂OH
:MFCD00004708